Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ALKBH5 Rabbit anti-Human, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma ABE547
Description
Detect ALKBH5 using this rabbit polyclonal antibody, Anti-ALKBH5 Antibody validated for use in western blotting and immunoprecipitation.
Specifications
ALKBH5 | |
Polyclonal | |
Purified rabbit polyclonal serum in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
Rabbit | |
Affinity Purified | |
Primary | |
Human |
Immunoprecipitation, Western Blot | |
Unconjugated | |
ALKBH5; ABH5; OFOXD1 | |
KLH-conjugated linear peptide corresponding to a region near the C-terminus of human ALKBH5. | |
100 μg | |
NP_060228.3 | |
2-8°C for 12 months |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction