Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

arachidonate 12-lipoxygenase, 12R type, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89004014 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-004-014 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-004-014 Supplier Abnova Corporation Supplier No. H00000242A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant ALOX12B.

This gene encodes an enzyme involved in the converstion of arachidonic acid to 12R-hydroxyeicosatetraenoic acid. Mutations in this gene are associated with nonbullous congenital ichthyosiform erythroderma. [provided by RefSeq

Sequence: NPNRPEWNGYIPGFPILINFKATKFLNLNLRYSFLKTASFFVRLGPMALAFKVRGLLDCKHSWKRLKDIRKIFPGKKSVVSEYVAEHWAED

Specifications

Antigen arachidonate 12-lipoxygenase, 12R type
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant ALOX12B.
Formulation 50% glycerol
Gene ALOX12B
Gene Accession No. NM_001139
Gene Alias 12R-LOX
Gene Symbols ALOX12B
Host Species Mouse
Immunogen ALOX12B (NP_001130, 171 a.a. ∼ 261 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Research Discipline Endocrine/Metabolism
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 242
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.