Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
alpha Actinin 2/3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA538605
Description
Alpha actinins belong to the spectrin gene superfamily which represents a diverse group of cytoskeletal proteins, including the alpha and beta spectrins and dystrophins. Alpha actinin is an actin-binding protein with multiple roles in different cell types. In nonmuscle cells, the cytoskeletal isoform is found along microfilament bundles and adherens-type junctions, where it is involved in binding actin to the membrane. In contrast, skeletal, cardiac, and smooth muscle isoforms are localized to the Z-disc and analogous dense bodies, where they help anchor the myofibrillar actin filaments. This gene encodes a muscle-specific, alpha actinin isoform that is expressed in both skeletal and cardiac muscles.
Specifications
alpha Actinin 2/3 | |
Polyclonal | |
Unconjugated | |
ACTN3 | |
CMD1AA, F-actin cross-linking protein, alpha-actinin skeletal muscle, alpha-actinin-2, Alpha-actinin-3, Alpha-actinin skeletal muscle isoform 3, ACTN3, alpha-Actinin3, Actinin alpha3, Actinin alpha3, alphaActinin-3, Actinin-alpha3, alpha-actinin-3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
11472, 11474, 171009, 291245, 88, 89 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P35609, Q08043, O88990, Q9JI91 | |
ACTN2, ACTN3 | |
A synthetic peptide derived from the N-terminal region of human Actin alpha-2/3 | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction