Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alpha Defensin-5 (HD5) Mouse anti-Human, Clone: 8C8, MilliporeSigma™

Catalog No. MABF31MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF31MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABF31MI Supplier MilliporeSigma Supplier No. MABF31
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Alpha Defensin-5 (HD5) Antibody, clone 8C8 detects level of Alpha Defensin-5 (HD5) and has been published and validated for use in immunohistochemistry and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Alpha Defensin-5 (HD5)
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 8C8
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG2bκ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_066290
Gene Symbols DEF5, DEFA5, HD-5
Host Species Mouse
Immunogen Recombinant protein corresponding to human alpha defensin-5 (HD5).
Purification Method Protein G Purified
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.