Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-alpha-Epithelial Cell Sodium Channel (20-42), Polyclonal

Catalog No. 324870100UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
32-487-0100UG 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 32-487-0100UG Supplier MilliporeSigma™ Supplier No. 324870100UG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects α-Epithelial Cell Sodium Channel (20-42) Clone: in Human, Rat samples, and it is validated for Immunoprecipitation

Epithelial sodium channels are amiloride-sensitive members of the degenerin/epithelial sodium channel (Deg/ENaC) superfamily of ion channels. ENaC in the kidney, lung, and colon plays a role in trans-epithelial Na+ and fluid balance. ENaC also mediates aldosterone-dependent sodium reabsorption in the distal nephron of the kidney, thus regulating blood pressure. ENaC is believed to be regulated through association with the cystic fibrosis transmembrane conductance regulator (CFTR) chloride ion channel.
TRUSTED_SUSTAINABILITY

Specifications

Antigen α-Epithelial Cell Sodium Channel (20-42)
Applications Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
Formulation In PBS, 1mg/mL BSA.
Host Species Rabbit
Immunogen a synthetic peptide [LMKGNKREEQGLGPEPAAPQQPT(C)] corresponding to amino acids 20-42 of human α-epithelial cell sodium channel (α-ENaC), conjugated to carrier protein
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the ∽97kDa glycosylatedα-ENaC protein.
Target Species Human, Rat
Content And Storage −20°C
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.