Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-alpha1-Antitrypsin Clone: TMF1#4B5,

Catalog No. 1782601MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
17-826-01MG 1 mg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-826-01MG Supplier MilliporeSigma™ Supplier No. 1782601MG

Mouse Monoclonal Antibody

Specifically detects α1-Antitrypsin Clone: TMF1#4B5 in Human samples, and it is validated for Enzyme Immunoassay, Radioimmunoassay

TRUSTED_SUSTAINABILITY

Specifications

Antigen α1-Antitrypsin
Applications ELISA, Radioimmune Assays (RIA)
Classification Monoclonal
Clone TMF1#4B5
Conjugate Unconjugated
Formulation In PBS, pH 7.4.
Host Species Mouse
Immunogen purified human α1-antitrypsin
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes α1-antitrypsin in human serum. Does not cross-react with α1-acid glycoprotein, serum albumin, or other serum proteins. Antibody Target Gene Symbol: SerpinA3 Target Synonym: AACT, ACT, ALPHA 1 ANTICHYMOTRYPSIN, ALPHA1 ACT, Alpha1-antitrypsin proteinase inhibitor, Anti-chymotrypsin, Contrapsin Like Protease Inhibitor Related, Contraspin-like protease inhibitor, CPi-26, Eb22/5, GIG24, GIG25, MGC88254, MGC93363, Serine protease inhibitor, Serpina3n, Spi-2.2, SPI2-2, Spi2/eb.4, SPI3, Spin2c, Spin3 Entrez Gene Name: serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 3
Target Species Human
Content And Storage −20°C
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.