Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STE20-related kinase adaptor beta, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89004665 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-004-665 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-004-665 Supplier Abnova Corporation Supplier No. H00055437A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant STRADB.

This gene encodes a protein that belongs to the serine/threonine protein kinase STE20 subfamily. One of the active site residues in the protein kinase domain of this protein is altered, and it is thus a pseudokinase. This protein is a component of a complex involved in the activation of serine/threonine kinase 11, a master kinase that regulates cell polarity and energy-generating metabolism. This complex regulates the relocation of this kinase from the nucleus to the cytoplasm, and it is essential for G1 cell cycle arrest mediated by this kinase. The protein encoded by this gene can also interact with the X chromosome-linked inhibitor of apoptosis protein, and this interaction enhances the anti-apoptotic activity of this protein via the JNK1 signal transduction pathway. Two pseudogenes, located on chromosomes 1 and 7, have been found for this gene. [provided by RefSeq

Sequence: MSLLDCFCTSRTQVESLRPEKQSETSIHQYLVDEPTLSWSRPSTRASEVLCSTNVSHYELQVEIGRGFDNLTSVHLARHTPTGTLVTIKITNLEN

Specifications

Antigen STE20-related kinase adaptor beta
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant STRADB.
Formulation 50% glycerol
Gene STRADB
Gene Accession No. NM_018571
Gene Alias ALS2CR2/CALS-21/ILPIP/ILPIPA/MGC102916/PAPK/PRO1038
Gene Symbols STRADB
Host Species Mouse
Immunogen STRADB (NP_061041, 1 a.a. ∼ 95 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Research Discipline Kinases
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 55437
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.