Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aly/REF Mouse anti-Human, Clone: 11G5, MilliporeSigma™

Catalog No. 051510MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-151-0MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-151-0MI Supplier MilliporeSigma Supplier No. 051510

Mouse Monoclonal Antibody

Anti-Aly/REF Antibody, clone 11G5 is a Mouse Monoclonal Antibody for detection of Aly/REF also known as Ally of AML-1 and LEF-1 or THO complex 4 and has been validated in western blot, immunocytochemistry, immunoprecipitation and immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Aly/REF
Applications Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 11G5
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1κ in buffer containing 0.1MTris-Glycine (pH 7.4, 150mM NaCl) with 0.05% sodium azide.
Gene Accession No. NP_005773.3
Gene Symbols ALY, BEF, Tho4, THOC4
Host Species Mouse
Immunogen Recombinant protein corresponding to human Aly/REF.
Purification Method Protein G purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.