Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AMACR Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. AB4235MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB4235MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB4235MI Supplier MilliporeSigma Supplier No. AB4235
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Detect AMACR using this Anti-AMACR Antibody validated for use in western blot, immunohistochemistry (paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen AMACR
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Peptide affinity purified antibody. Liquid in PBS, with 0.02% sodium azide as a preservative.
Gene Symbols AMACR; RACE; EC 5.1.99.4
Host Species Rabbit
Immunogen Synthetic human AMACR peptide.
Purification Method Affinity Purified
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_014324.4; NM_203382.1
Target Species Human
Content And Storage 2-8°C in undiluted aliquots for 3 months or −20°C for up to 12 months. Avoid repeated freeze-thaw cycles.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.