Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AMBN, Rabbit, Monoclonal Antibody, Abnova™

Catalog No. 89206299 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg x up to 3 Abs
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-206-299 1 mg x up to 3 Abs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-206-299 Supplier Abnova Corporation Supplier No. H00000258K

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Includes

Up to three rabbit IgG clones of 1mg each will be delivered to customer.

Rabbit monoclonal antibody raised against a human AMBN peptide using ARM Technology.

Ameloblastin is thought to represent an unique ameloblast-specific gene product that may be important in enamel matrix formation and mineralization. The gene is located on chromosome 4 near other genes associated with mineralized tissues: osteopontin, bone sialoprotein, and bone morphogenetic protein 3. Based on its cytogenetic location, this gene is a candidate gene for one form of the disorder, dentinogenesis imperfecta, and/or the disorder, autosomal dominant amylogenesis imperfecta. [provided by RefSeq

Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)
Expression: Overexpression vector and transfection into 293H cell line

Order Info

  1. Customer may provide cell or tissue lysate for antibody screening.
  2. Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.

Specifications

Antigen AMBN
Applications ELISA, Western Blot
Classification Monoclonal
Concentration 1mg
Conjugate Unconjugated
Description ameloblastin (enamel matrix protein)
Formulation In 1X PBS, pH 7.4
Gene AMBN
Gene Accession No. AMBN
Gene Symbols AMBN
Host Species Rabbit
Immunogen A synthetic peptide of human AMBN
Quantity 1 mg x up to 3 Abs
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 258
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.