Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AMBP, Mouse, Clone: 4A7, Abnova™

Catalog No. 89016130 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-016-130 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-016-130 Supplier Abnova Corporation Supplier No. H00000259M09
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length native AMBP.

This gene encodes a complex glycoprotein secreted in plasma. The precursor is proteolytically processed into distinct functioning proteins: alpha-1-microglobulin, which belongs to the superfamily of lipocalin transport proteins and may play a role in the regulation of inflammatory processes, and bikunin, which is a urinary trypsin inhibitor belonging to the superfamily of Kunitz-type protease inhibitors and plays an important role in many physiological and pathological processes. This gene is located on chromosome 9 in a cluster of lipocalin genes. [provided by RefSeq

Specifications

Antigen AMBP
Applications ELISA, Immunofluorescence
Classification Monoclonal
Clone 4A7
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full length native AMBP.
Dilution The optimal working dilution should be determined by the end user.
Formulation PBS with no preservative; pH 7.4
Gene AMBP
Gene Accession No. P02760
Gene Alias EDC1/HCP/HI30/IATIL/ITI/ITIL/ITILC/UTI
Gene Symbols AMBP
Host Species Mouse
Immunogen Native purified human AMBP.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 259
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.