Learn More
amelogenin (amelogenesis imperfecta 1, X-linked), Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human AMELX protein.
Supplier: Abnova Corporation H00000265D01P
Description
This gene encodes a member of the amelogenin family of extracellular matrix proteins. Amelogenins are involved in biomineralization during tooth enamel development. Mutations in this gene cause X-linked amelogenesis imperfecta. Alternative splicing results in multiple transcript variants encoding different isoforms. [provided by RefSeq
Sequence: MGTWILFACLLGAAFAMPLPPHPGHPGYINFSYEVLTPLKWYQSIRPPYPSYGYEPMGGWLHHQIIPVLSQQHPPTHTLQPHHHIPVVPAQQPVIPQQPMMPVPGQHSMTPIQHHQPNLPPPAQQPYQPQPVQPQPHQPMQPQPPVHPMQPLPPQPPLPPMFPMQPLPPMLPDLTLEAWPSTDKTKREEVDSpecifications
amelogenin (amelogenesis imperfecta 1, X-linked) | |
Polyclonal | |
Rabbit polyclonal antibody raised against a full-length human AMELX protein. | |
AMELX | |
AIH1/ALGN/AMG/AMGL/AMGX | |
Rabbit | |
Affinity Purified | |
RUO | |
265 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
Western Blot | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_001142.2 | |
AMELX | |
AMELX (NP_001133.1, 1 a.a. ∼ 191 a.a) full-length human protein. | |
100 μg | |
Primary | |
Human | |
Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.