Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AMHR2 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA513901
Description
This antibody is predicted to react with mouse based on sequence homology.
The AMH receptor (AMHR or AMHR2) is a serine/threonine kinase with a single transmembrane domain belonging to the family of type II receptors for TGF-beta-related proteins. Anti-Mullerian hormone (AMH) and its receptor are involved in the regression of Mullerian ducts in male fetuses. Male sex differentiation is mediated by 2 discrete hormones produced by the fetal testis. Testosterone, produced by Leydig cells, virilizes the external genitalia and promotes prostatic growth; anti-Mullerian hormone (AMH) results in regression of Mullerian ducts which would otherwise differentiate into the uterus and fallopian tubes.
Specifications
AMHR2 | |
Polyclonal | |
Unconjugated | |
Q16671 | |
AMHR2 | |
KLH conjugated synthetic peptide between 65-91 amino acids from the N-terminal region of human AMHR2 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
0.35 mg/mL | |
AMHR2 | |
Anti-Muellerian hormone type II receptor [Precursor]; AMH type II receptor; MIS type II receptor; MISRII; MRII; AMHR2; AMHR; | |
Rabbit | |
Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
RUO | |
269 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction