Learn More
AMOT, Rabbit, Monoclonal Antibody, Abnova™
Rabbit monoclonal antibody raised against a human AMOT peptide using ARM Technology.
Supplier: Abnova Corporation H00154796K
Includes: Up to three rabbit IgG clones of 1mg each will be delivered to customer.
Description
This gene belongs to the motin family of angiostatin binding proteins characterized by conserved coiled-coil domains and C-terminal PDZ binding motifs. The encoded protein is expressed predominantly in endothelial cells of capillaries as well as larger vessels of the placenta where it may mediate the inhibitory effect of angiostatin on tube formation and the migration of endothelial cells toward growth factors during the formation of new blood vessels. Alternative splicing results in multiple transcript variants encoding different isoforms. [provided by RefSeq
Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)Expression: Overexpression vector and transfection into 293H cell line
Order Info
- Customer may provide cell or tissue lysate for antibody screening.
- Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.
Specifications
AMOT | |
Monoclonal | |
Unconjugated | |
In 1X PBS, pH 7.4 | |
AMOT | |
AMOT | |
A synthetic peptide of human AMOT | |
RUO | |
154796 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
ELISA, Western Blot | |
1mg | |
angiomotin | |
AMOT | |
KIAA1071 | |
Rabbit | |
1 mg x up to 3 Abs | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.