Learn More
Description
Adenosine monophosphate deaminase-2 (EC 3.5.4.6) catalyzes the deamination of AMP to IMP and plays an important role in the purine nucleotide cycle.[supplied by OMIM
Specifications
Specifications
| Antigen | AMPD2 |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 2F5 |
| Conjugate | Unconjugated |
| Description | Mouse monoclonal antibody raised against a partial recombinant AMPD2. |
| Formulation | ascites with no preservative |
| Gene | AMPD2 |
| Gene Accession No. | NM_139156 |
| Gene Symbols | AMPD2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.