Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

amphiphysin, Mouse, Clone: 1H3-1D11, Abnova™

Catalog No. 89001423 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-001-423 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-001-423 Supplier Abnova Corporation Supplier No. H00000273M01A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant AMPH.

This gene encodes a protein associated with the cytoplasmic surface of synaptic vesicles. A subset of patients with stiff-man syndrome who were also affected by breast cancer are positive for autoantibodies against this protein. Alternate splicing of this gene results in two transcript variants encoding different isoforms. Additional splice variants have been described, but their full length sequences have not been determined. [provided by RefSeq

Sequence: MADIKTGIFAKNVQKRLNRAQEKVLQKLGKADETKDEQFEEYVQNFKRQEAEGTRLQRELRGYLAAIKGMQEASMKLTESLHEVYEPDWYGREDVKMVGEKCDVLWEDFHQKLVDGSLLTLDTYLGQFPDIKNRIAKRSRKLVDYDSARHHLEALQSSKRKDESRISKAEEEFQKAQKVFEEFNVDLQEELPSLWSRRVGFYVNTFKNVSSLEAKFHKEIAVLCHKLYEVMTKLGDQHADKAFTIQGAPSDSGPLRIAKTPSPPEEPSPLPSPTASPNHTLAPASPAPARPRSPSQTRKGPPVPPLPKVTPTKELQQENIISFFEDNFVPEISVTTPSQNEVPEVKKEETLLDLDFDPFKPEVTPAGSAGVTHSPMSQTLPWDLWTTSTDLVQPASGGSFNGFTQPQDTSLFTMQTDQSMICNLAESEQAPPTEPKAEEPLAAVTPAVGLDLGMDTRAEEPVEEAVIIPGADADAAVGTLVSAAEGAPGEEAEAETATVPAGEGVSLEEAKIGTETTEGAESAQPEAEELEATVPQEKVIPSVVIEPASNHEEEGENEITIGAEPKETTEDAAPPGPTSETPELATEQKPIQDPQPTPSAPAMGAADQLASAREASQELPPGFLYKVETLHDFEAANSDELTLQRGDVVLVVPSDSEADQDAGWLVGVKESDWLQYRDLATYKGLFPENFTRRLD

Specifications

Antigen amphiphysin
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1H3-1D11
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full length recombinant AMPH.
Formulation ascites with no preservative
Gene AMPH
Gene Accession No. BC034376
Gene Alias AMPH1
Gene Symbols AMPH
Host Species Mouse
Immunogen AMPH (AAH34376.1, 1 a.a. ∼ 695 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 273
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.