Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amphiphysin II Rabbit anti-Mouse, Rat, Polyclonal, MilliporeSigma™

Catalog No. ABT21MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABT21MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABT21MI Supplier MilliporeSigma Supplier No. ABT21
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Detect Amphiphysin II using this Anti-Amphiphysin II Antibody validated for use in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Amphiphysin II
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4, 150mM NaCl) with 0.05% sodium azide.
Gene Symbols AMPHL; AMPH2; BIN1; SH3P9
Host Species Rabbit
Immunogen KLH-conjugated recombinant protein corresponding to human Amphiphysin II.
Purification Method Affinity Purified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NP_004296
Target Species Mouse, Rat
Content And Storage 2-8°C for 12 months
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.