Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amphiregulin Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™

Catalog No. AB3152MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB3152MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB3152MI Supplier MilliporeSigma Supplier No. AB3152
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This Anti-Amphiregulin Antibody is validated for use in ELISA, immunoprecipitation, western blotting for the detection of Amphiregulin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Amphiregulin
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified immunoglobulin from rabbit anti-serum by Protein A chromatography. Liquid in 10mM PBS, pH 7.4, with 0.2% BSA and 15mM sodium azide.
Gene Symbols AREG; MGC13647; CRDGF; amphiregulin; AR; SDGF
Host Species Rabbit
Immunogen A synthetic 19-mer peptide, corresponding to aa 26-44 from the secreted amphiregulin of human origin.
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_001657.2
Target Species Human, Mouse
Content And Storage 2-8°C in undiluted aliquots for up to 12 months
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.