Learn More
AMY2B, Mouse, Clone: 2B12, Abnova™
Mouse monoclonal antibody raised against a partial recombinant AMY2B.
Supplier: Abnova Corporation H00000280M01
Description
Amylases are secreted proteins that hydrolyze 1,4-alpha-glucoside bonds in oligosaccharides and polysaccharides, and thus catalyze the first step in digestion of dietary starch and glycogen. The human genome has a cluster of several amylase genes that are expressed at high levels in either salivary gland or pancreas. This gene encodes an amylase isoenzyme produced by the pancreas. [provided by RefSeq
Sequence: PNTQQGRTSIVHLFEWRWVDIALECERYLAPKGFGGVQVSPPNENVAIHNPFRPWWERYQPVSYKLCTRSGNEDEFRNMVTRCNNVGVRIYVDAVINHMSpecifications
AMY2B | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_020978 | |
AMY2B | |
AMY2B (NP_066188, 19 a.a. ∼ 117 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a κ |
ELISA | |
2B12 | |
Mouse monoclonal antibody raised against a partial recombinant AMY2B. | |
AMY2B | |
AMY2 | |
Mouse | |
Affinity chromatography | |
RUO | |
280 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.