Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amyloid Β42 Mouse anti-Human, Mouse, Clone: G2-11, MilliporeSigma™

Catalog No. MABN12MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABN12MI 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABN12MI Supplier MilliporeSigma Supplier No. MABN12
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Amyloid β42 Antibody, clone G2-11 detects level of Amyloid β42 and has been published and validated for use in western blot, western blot, ELISA, immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Amyloid β42
Applications ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone G2-11
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1κ in buffer containing 0.1MTris-Glycine (pH 7.4, 150mM NaCl) with 0.05% sodium azide.
Gene Accession No. NP_000475
Gene Symbols APP, A4, AAA, ABETA, ABPP, AD1, APPI, CTFgamma, CVAP, PN-II, PN2, PreA4
Host Species Mouse
Immunogen KLH-conjugated linear peptide corresponding to the C-terminus of Amyloid β42.
Purification Method Protein G purfied
Quantity 50 μg
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.