Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amyloid Mouse anti-Bovine, Human, Monkey, Clone: AB10, MilliporeSigma™

Catalog No. MAB5208100U Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB5208100U 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB5208100U Supplier MilliporeSigma Supplier No. MAB5208100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Amyloid Antibody, β 1-16, clone AB10 detects level of Amyloid and has been published and validated for use in ELISA, immunoprecipitation, western blot, immunocytochemistry, immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Amyloid
Applications ELISA, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone AB10
Conjugate Unconjugated
Formulation Purified immunoglobulin. Liquid in TRIS-buffered saline containing 1mg/mL bovine serum albumin and 0.01% sodium azide.
Gene Accession No. NM_000484.2; NM_201413.1; NM_201414.1
Gene Symbols APP, ABPP, PN-II, AD1, A4, CVAP, PN2, ABETA, CTFgamma, APPI, PreA4, AAA
Host Species Mouse
Immunogen Amyloid-beta 1-16 conjugated to KLH
Quantity 100 μg
Primary or Secondary Primary
Target Species Bovine, Human, Monkey
Content And Storage −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.