Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Amyloid Fibrils LOC Rabbit anti-Human, Polyclonal

Catalog No. AB2287MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB2287MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB2287MI Supplier MilliporeSigma™ Supplier No. AB2287
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This Anti-Amyloid Fibrils LOC Antibody is validated for use in immunoprecipitation, immunocytochemistry, immunohistochemistry, ELISA, western blot, DB for the detection of Amyloid Fibrils LOC.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Amyloid Fibrils LOC
Applications Dot Blot, ELISA, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Unpurified rabbit polyclonal antibody serum containing 0.05% sodium azide.
Gene Symbols Amyloid fibrils LOC
Host Species Rabbit
Immunogen Fibrils prepared from human islet amyloid polypeptide.
Purification Method Unpurified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_000484.2; NM_201413.1; NM_201414.1
Target Species Human
Content And Storage -20°C in undiluted aliquots for up to 12 months, Avoid Freeze/Thaw Cycles
Form Serum
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.