Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amyloid Fibrils OC Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. AB2286MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB2286MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB2286MI Supplier MilliporeSigma Supplier No. AB2286
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Amyloid Fibrils OC Antibody is an antibody against Amyloid Fibrils OC for use in immunoprecipitation, immunocytochemistry, immunohistochemistry, ELISA, western blot, DB.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Amyloid Fibrils OC
Applications Dot Blot, ELISA, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Unpurified rabbit polyclonal antibody serum containing 0.05% sodium azide.
Gene Symbols Amyloid Fibrils OC
Host Species Rabbit
Immunogen Fibrils prepared from human Aβ42 peptide.
Purification Method Unpurified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_000484.2; NM_201413.1; NM_201414.1
Target Species Human
Content And Storage -20°C in undiluted aliquots for up to 12 months, Avoid Freeze/Thaw Cycles
Form Serum
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.