Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amyloid Β Precursor Protein Mouse anti-Human, Clone: 4.1, MilliporeSigma™

Catalog No. MAB5354MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB5354MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB5354MI Supplier MilliporeSigma Supplier No. MAB5354
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Amyloid β Precursor Protein Antibody, KPI domain, clone 4.1 detects level of Amyloid β Precursor Protein and has been published and validated for use in ELISA, western blot, immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Amyloid β Precursor Protein
Applications ELISA, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 4.1
Conjugate Unconjugated
Formulation Purified immunoglobulin. Liquid in 20mM Sodium phosphate buffer with 150mM NaCl.
Gene Accession No. NM_000484.2; NM_201413.1; NM_201414.1
Gene Symbols APP, ABPP, PN-II, AD1, A4, CVAP, PN2, ABETA, CTFgamma, APPI, PreA4, AAA
Host Species Mouse
Immunogen Purified recombinant KPI domain of Amyloid-beta precursor protein.
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C in undiluted aliquots for up to 6 months.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.