Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amyloid Precursor Protein Rabbit anti-Human, Monkey, Polyclonal, MilliporeSigma™

Catalog No. AB5988PMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB5988PMI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB5988PMI Supplier MilliporeSigma Supplier No. AB5988P

Rabbit Polyclonal Antibody

Anti-Amyloid Precursor Protein Antibody, Domain 770 detects level of Amyloid Precursor Protein and has been published and validated for use in immunoprecipitation, western blot, immunocytochemistry, immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Amyloid Precursor Protein
Applications Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Affinity purified immunoglobulin. Liquid in TRIS-buffered saline with 0.01% sodium azide.
Gene Symbols APP; ABPP; PN-II; AD1; A4; CVAP; PN2; ABETA; CTFgamma; APPI; PreA4; AAA
Host Species Rabbit
Immunogen Synthetic peptide corresponding to the OX2 domain of APP770.
Purification Method Affinity Purified
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_000484.2; NM_201413.1; NM_201414.1
Target Species Human, Monkey
Content And Storage 2-8°C in undiluted aliquots for up to 6 months
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.