Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Androgen Receptor Mouse anti-Human, Unlabeled, Clone: G122-25, BD

Catalog No. BDB554224 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB554224 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB554224 Supplier BD Biosciences Supplier No. 554224
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The androgen receptor (AR) mediating androgen effects is a member of the steroid and thyroid hormone receptor gene superfamily encoding ligand-dependent nuclear transcription factors. They can be divided into three main domains, an N-terminal domain which modulates transcription efficiency, a central DNA domain which binds to a target gene hormone response element, and a C-terminal hormone binding domain. Androgens are necessary for normal male development and function. They exert their effects on target tissue through binding to the AR, followed by association of the AR complex with specific binding sites on DNA. This AR complex may induce or repress gene transcription. Mutations in the AR gene are associated with androgen insensitivity syndrome, a disorder that causes XY genotypic males to develop as phenotypic females because of their inability to respond to androgens. AR mutations have also been identified in human prostate cancer specimens and the LNCaP human prostate cancer cell line. Monoclonal and polyclonal antibodies to the AR have identified AR-positive cells in a variety of tissues including male and female sexual organs, kidney, liver, adrenal cortex, pituitary gland, skeletal, cardiac, and smooth muscle cells. The androgen receptor migrates at a molecular weight of ∽100 kD in SDS-PAGE. G122–25 recognizes an epitope localized within the DNA-binding domain (between amino acids 486 and 651) of the human androgen receptor. Additionally, G122–25 has been shown to recognize androgen receptors in frozen rat prostate tissue sections. G122–25 does not cross-react with estrogen or progesterone receptors. Purified recombinant human androgen receptor protein was used as immunogen.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Androgen Receptor
Applications Immunohistochemistry
Classification Monoclonal
Clone G122-25
Concentration 0.5mg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Purified human androgen receptor recombinant protein
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human, Rat
Content And Storage Store undiluted at 4°C
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.