Learn More
MilliporeSigma™ anti-Angiopoietin-1, N-terminus (Ang-1); Polyclonal; ™ (Chemicon™)
Supplier: MilliporeSigma™ AB3120/DEL

Description
The remodelling of the primitive vasculature in mammals to form the mature cardiovascular system includes a number of factors directing endothelial cell development. Angiopoietin-1 (mouse and human Ang-1; 498aa with 98% identity) is an angiogenic secreted protein that interacts with the endothelial specific Tie-2 receptor. It is primarily expressed in developing endothelial cells. During embryonic dvevelopment, Ang-1 binds and induces tyrosine phosphorylation of Tie-2. Ang-1 appears to play a crucial role in mediating matrix and mesenchyme. It mediates blood vessel maturation and stability. Ang-1 may play a critical role in heart development. Ang-1 deficient mice mimic the phenotype exhibited by animals deficient in Tie-2. A homolog of Ang-1, termed Angiopoietin-2 (mouse and human Ang-2, 496aa with 85% identity) may act as an antagonist for Ang-1 and Tie-2. Ang-1 and Ang-2 have ∽60% sequence homology.
ELISA, Western Blotting

Specifications
Angiopoietin-1, N-terminus (Ang-1) | |
Unconjugated | |
20 amino acid peptide sHuence near the amino-terminus of mouse Ang-1. | |
50 μg | |
Cell Structure | |
Mouse |
Polyclonal | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.