Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Angiopoietin-1 Rabbit anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™

Catalog No. AB10516MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB10516MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB10516MI Supplier MilliporeSigma Supplier No. AB10516
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Angiopoietin-1 Antibody is an antibody against Angiopoietin-1 for use in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Angiopoietin-1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Rabbit polyclonal serum in buffer containing 0.1M Tris-Glycine (pH7.4) 150mM NaCl with 0.05% sodium azide.
Gene Symbols ANGPTL2; ARP4; FIAF; HFARP; NL2; PGAR; pp1158
Host Species Rabbit
Immunogen This antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide selected from mouse Angiopoietin-1.
Purification Method Antigen affinity purified
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_001146.3
Target Species Human, Mouse, Rat
Content And Storage 2-8°C for 12 months
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.