Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Angiotensin Converting Enzyme Mouse anti-Human, Hamster, Monkey, Rat, Feline, Clone: 9B9, MilliporeSigma™

Catalog No. MAB4051MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB4051MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB4051MI Supplier MilliporeSigma Supplier No. MAB4051
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Angiotensin Converting Enzyme Antibody, clone 9B9 detects level of Angiotensin Converting Enzyme and has been published and validated for use in ELISA, immunoprecipitation, western blot, immunocytochemistry, immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Angiotensin Converting Enzyme
Applications ELISA, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 9B9
Conjugate Unconjugated
Formulation Purified immunoglobulin. Liquid in PBS, pH 7.4, 150mM NaCl containing 0.05% sodium azide.
Gene Accession No. NM_152831.1; NM_000789.2; NM_152830.1
Gene Symbols ACE, DCP, ACE1, carboxycathepsin, ACE-T, MGC26566, DCP1, CD143, EC 3.4.15.1, EC 3.2.1.- [Contains: Angiotensin-converting enzyme, testis- specific isoform, soluble form]., EC 3.4.15.1 [Contains: Angiotensin-converting enzyme, somatic isoform, soluble form
Host Species Mouse
Immunogen Angiotensin-converting enzyme from human lung
Quantity 100 μg
Primary or Secondary Primary
Target Species Feline, Hamster, Human, Monkey, Rat
Content And Storage −20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.