Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Angiotensin-converting enzyme Rabbit anti-Human, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma ABC246
Description
Detect Angiotensin Converting Enzyme (ACE) using this rabbit polyclonal antibody, Anti-Angiotensin-converting enzyme Antibody validated for use in western blotting and immunohistochemistry (Paraffin).
Specifications
Angiotensin-converting enzyme | |
Polyclonal | |
Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
Rabbit | |
Affinity Purified | |
Primary | |
Human |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
ACE; DCP; DCP1 | |
KLH-conjugated linear peptide corresponding to the extracellular domain of human Angiotensin-converting enzyme. | |
100 μL | |
NP_000780 | |
2-8°C for 12 months |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction