Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ANGPTL4 (MID) Rabbit anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™

Catalog No. AB10605MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB10605MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB10605MI Supplier MilliporeSigma Supplier No. AB10605
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-ANGPTL4 (MID) Antibody is an antibody against ANGPTL4 (MID) for use in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ANGPTL4 (MID)
Applications Western Blot
Classification Polyclonal
Clone ZMD.521
Conjugate Unconjugated
Formulation 400μL aliquot at a concentration of 0.25mg/mL in phosphate buffered saline (pH 7.4) containing 0.1% sodium azide.
Gene Symbols ANGPTL4; UNQ171/PRO197; HFARP; ANGPTL2; PSEC0166; PGAR; NL2; pp1158; ARP4; FIAF
Host Species Rabbit
Immunogen Synthetic peptide derived from an internal region of the human ANGTPL4 protein, which differs from rat and mouse by one and two conservative amino acid replacements, respectively.
Purification Method Immunoaffinity purified
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_001039667.1; NM_139314.1
Target Species Human, Mouse, Rat
Content And Storage −20°C in undiluted aliquotes for 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.