Learn More
Description
Specifications
Specifications
| Antigen | Anion Exchanger 1 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Affinity purified immunoglobulin. Liquid in Liquid in PBS with 0.1% BSA. |
| Gene Symbols | SLC4A1; Wr; MGC126619; MGC116750; MGC126623; BND3; MGC116753; DI; CD233; WD; WD1; EMPB3; AE1; EPB3; SW; RTA1A; FR; WR |
| Host Species | Rabbit |
| Immunogen | A 20 amino acid peptide sequence near the N-terminus of rat AE1 (Kudrycki et al. 1989; Kopito et al. 1985; Raida et al. 1989; Lux et al. 1989; Tanner et al. 1988). |
| Purification Method | Affinity Purified |
| Quantity | 50 μg |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.