Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ankyrin Mouse anti-Bovine, Canine, Human, Mouse, Pig, Rat, Rabbit, Clone: Ank016, MilliporeSigma™

Catalog No. MAB1683MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1683MI 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB1683MI Supplier MilliporeSigma Supplier No. MAB1683
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Ankyrin Antibody, clone Ank016 is an antibody against Ankyrin for use in ELISA, flow cytometry, immunoprecipitation, western blotting and immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ankyrin
Applications ELISA, Flow Cytometry, Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone Ank016
Conjugate Unconjugated
Formulation Purified immunoglobulin from culture supernatant. Liquid in 10mM sodium phosphate, pH 7.5 and 150mM NaCl with 0.01% NaN3.
Gene Accession No. NM_020479.2; NM_020481.2; NM_000037.3; NM_020475.2; NM_020476.2; NM_020477.2; NM_020478.3; NM_020480.3
Gene Symbols ANK1, ANK, SPH2, SPH1, Ankyrin-R, Ankyrin-1, ankyrin-R
Host Species Mouse
Immunogen Purified human erythroid cells ankyrin
Quantity 50 μg
Primary or Secondary Primary
Target Species Bovine, Canine, Human, Mouse, Pig, Rabbit, Rat
Content And Storage −20°C in undiluted aliquots for 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.