Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ankyrin-G Mouse anti-Rat, Clone: N106/36, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MABN466
Description
Anti-Ankyrin-G Antibody, clone N106/36 is a highly specific mouse monoclonal antibody and that targets Ankyrin and has been tested in immunohistochemistry.
Specifications
Ankyrin-G | |
Monoclonal | |
Unconjugated | |
NP_066267 | |
Mouse | |
Protein G purified | |
Primary | |
2-8°C for one year | |
IgG1 κ |
Immunohistochemistry | |
N106/36 | |
Purified mouse monoclonal IgG1κ in 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
ANK3 | |
Recombinant protein corresponding to human Ankyrin-G. | |
100 μg | |
Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction