Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ANKRD45 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA521796
Description
Recommended positive controls: HepG2. Predicted reactivity: Mouse (81%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
ANKRD45, or Ankyrin Repeat Domain 45, belongs to the large family of ankyrin repeat domain-containing proteins. Proteins with ankyrin repeats typically serve as scaffolds for protein-protein interactions, thus playing a critical role in various cellular processes, such as signal transduction, cell cycle regulation, and the maintenance of cellular structure. While the specific functions of ANKRD45 remain largely unexplored, studies on related ankyrin repeat proteins suggest that they may have roles in cellular stress responses and cancer development.
Specifications
ANKRD45 | |
Polyclonal | |
Unconjugated | |
ANKRD45 | |
4933409K03Rik; ANKRD45; ankyrin repeat domain 45; ankyrin repeat domain-containing protein 45; cancer/testis antigen 117; CT117; RGD1311153; RP3-436N22.4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
339416 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
Q5TZF3 | |
ANKRD45 | |
Recombinant protein encompassing a sequence within the center region of human ANKRD45. The exact sequence is proprietary. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction