Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Annexin I Mouse, Unlabeled, Clone: 29, BD

Catalog No. BDB610066 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610066 50 μg
BDB610067 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB610066 Supplier BD Biosciences Supplier No. 610066
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Annexin I, also known as lipocortin-1 or calpactin II, is part of a calcium-dependent phospholipid-binding protein family. Members share a common core domain, but each has a unique N-terminal tail that imparts its functional specificity. It is believed that phosphorlyation of this region in annexin I regulates its functions. Annexin I has been implicated in regulating phospholipid vesicle aggregation, mediating inflammatory response, and inhibiting the activity of phospholipase A2. Additionally, it has been found that the 10kDa protein S100C influences annexin I's activity by binding to the N-terminal 13 residues of annexin in a manner similar to that of the annexin II/p11 complex.

Host Species: Mouse
Clone: 29
Isotype: IgG1
Species Reactivity: Human
Immunogen: Cow Annexin I aa. 1-346
Formula Weight [Chemical]: 38kDa

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Annexin I
Applications Western Blot
Classification Monoclonal
Clone 29
Concentration 250μg/mL
Conjugate Unconjugated
Description Lipocortin-1; Calpactin II
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Cow Annexin I aa. 1-346
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Human
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.