Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Annexin II Mouse, Unlabeled, Clone: 5, BD

Catalog No. BDB610068 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610068 50 μg
BDB610069 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB610068 Supplier BD Biosciences Supplier No. 610068
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Annexin II (p36), originally discovered as a substrate for the src oncogene, is one of the most studied members of the annexin family of calcium-dependent phospholipid-binding proteins. It exists in cells as either a monomer or a heterotetramer complexed with p11, an S100-related protein. In vitro, it has been found that unphosphorylated annexin II is capable of causing aggregation of chromaffin granules, and, upon phosphorylation, induces fusion of the granules membranes. A similiar phenomenon may occur in the final steps of exocytosis after cytoplasmic annexin translocates to the subplasmalemmal region and is phosphorylated by protein kinase C. However, other studies refute annexin II's involvement in exocytosis, and a conclusive role for annexin II in membrane trafficking remains debated.

Host Species: Mouse
Clone: 5
Isotype: IgG1
Species Reactivity: Human
Immunogen: Human Annexin II aa. 123-328
Formula Weight [Chemical]: 36kDa

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Annexin II
Applications Western Blot
Classification Monoclonal
Clone 5
Concentration 250μg/mL
Conjugate Unconjugated
Description Lipocortin II, Calpactin I
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human Annexin II aa. 123-328
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Chicken, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.