Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Annexin II Light Chain Mouse, Unlabeled, Clone: 148, BD

Catalog No. BDB610070 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610070 50 μg
BDB610071 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB610070 Supplier BD Biosciences Supplier No. 610070
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Annexin II often exists as a heterotetramer complexed with an 11kDa light chain. This p11 subunit is related to the S100 family of calcium-binding proteins but mutations have rendered it unable to bind calcium. The p11 protein binds to the amino terminus of the annexin II monomer, but not to other annexins. It may regulate the function of annexin II, as the tetrameric form seems to have a higher calcium and phospholipid-binding affinity than the monomeric annexin II. In the cell, p11 has been localized to the cytoplasmic face of the plasma membrane where the annexin II tetramer is known to exist.

Host Species: Mouse
Clone: 148
Isotype: IgG1
Species Reactivity: Human
Immunogen: Cow Annexin II Light Chain aa. 1-97
Formula Weight [Chemical]: 11kDa

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Annexin II Light Chain
Applications Western Blot
Classification Monoclonal
Clone 148
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Cow Annexin II Light Chain aa. 1-97
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Chicken, Human
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.