Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Annexin VI Mouse anti-Human, Mouse, Clone: 4B11.1, MilliporeSigma™

Catalog No. MABC272MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABC272MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABC272MI Supplier MilliporeSigma Supplier No. MABC272
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-Annexin VI Antibody, clone 4B11.1 is validated for use in western blotting and immunocytochemistry for the detection of Annexin VI.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Annexin VI
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 4B11.1
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG2aκ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_001146
Gene Symbols ANXA6, ANX6
Host Species Mouse
Immunogen GST-tagged recombinant protein corresponding to human Annexin VI.
Purification Method Protein G purified
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.