Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Anti-Desmin Mouse, Unlabeled, Clone: RD301, BD

Catalog No. BDB550626 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB550626 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB550626 Supplier BD Biosciences Supplier No. 550626

Mouse Monoclonal Antibody

Intermediate filaments (IF) are a subset of cytoskeletal proteins which function to give overall structural integrity to the plasma membrane as well as organize cells into specific tissues. IF proteins can be divided into six major types based upon the similarity in sequence. Desmin belongs to the type III category of IF proteins which are predominantly expressed in muscle cells including cardiac, skeletal and smooth muscle. Furthermore, the expression of desmin is regulated in a stage and tissue-specific manner, since it is induced during terminal differentiation of skeletal muscle cells. In skeletal cardiac muscle cells, desmin is localized in the Z-disk region and at the intercalated disk and acts to stabilize sarcomeres in stimulated muscle. Desmin migrates in SDS/PAGE as a 53 kDa protein.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Anti-Desmin
Applications Western Blot
Classification Monoclonal
Clone RD301
Concentration 0.5mg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Purified desmin from chicken gizzard
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Chicken, Human, Mouse, Rat
Content And Storage Store undiluted at 4°C
Isotype IgG2b
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.