Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

annexin A4, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89017998 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-017-998 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-017-998 Supplier Abnova Corporation Supplier No. H00000307D01P
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against a full-length human ANXA4 protein.

Annexin IV (ANX4) belongs to the annexin family of calcium-dependent phospholipid binding proteins. Although their functions are still not clearly defined, several members of the annexin family have been implicated in membrane-related events along exocytotic and endocytotic pathways. ANX4 has 45 to 59% identity with other members of its family and shares a similar size and exon-intron organization. Isolated from human placenta, ANX4 encodes a protein that has possible interactions with ATP, and has in vitro anticoagulant activity and also inhibits phospholipase A2 activity. ANX4 is almost exclusively expressed in epithelial cells. [provided by RefSeq

Sequence: MAMATKGGTVKAASGFNAMEDAQTLRKAMKGLGTDEDAIISVLAYRNTAQRQEIRTAYKSTIGRDLIDDLKSELSGNFEQVIVGMMTPTVLYDVQELRRAMKGAGTDEGCLIEILASRTPEEIRRISQTYQQQYGRSLEDDIRSDTSFMFQRVLVSLSAGGRDEGNYLDDALVRQDAQDLYEAGEKKWGTDEVKFLTVLCSRNRNHLLHVFDEYKRISQKDIEQSIKSETSGSFEDALLAIVKCMRNKSAYFAEKLYKSMKGLGTDDNTLIRVMVSRAEIDMLDIRAHFKRLYGKSLYSFIKGDTSGDYRKVLLVLCGGDD

Specifications

Antigen annexin A4
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against a full-length human ANXA4 protein.
Formulation PBS with no preservative; pH 7.4
Gene ANXA4
Gene Accession No. NM_001153.2
Gene Alias ANX4/DKFZp686H02120/MGC75105/PIG28/ZAP36
Gene Symbols ANXA4
Host Species Rabbit
Immunogen ANXA4 (NP_001144.1, 1 a.a. ∼ 321 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 307
Target Species Human, Mouse
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.