Learn More
annexin A5, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human ANXA5 protein.
Supplier: Abnova Corporation H00000308D01P
Description
The protein encoded by this gene belongs to the annexin family of calcium-dependent phospholipid binding proteins some of which have been implicated in membrane-related events along exocytotic and endocytotic pathways. Annexin 5 is a phospholipase A2 and protein kinase C inhibitory protein with calcium channel activity and a potential role in cellular signal transduction, inflammation, growth and differentiation. Annexin 5 has also been described as placental anticoagulant protein I, vascular anticoagulant-alpha, endonexin II, lipocortin V, placental protein 4 and anchorin CII. The gene spans 29kb containing 13 exons, and encodes a single transcript of approximately 1.6kb and a protein product with a molecular weight of about 35kDa. [provided by RefSeq
Sequence: MAQVLRGTVTDFPGFDERADAETLRKAMKGLGTDEESILTLLTSRSNAQRQEISAAFKTLFGRDLLDDLKSELTGKFEKLIVALMKPSRLYDAYELKHALKGAGTNEKVLTEIIASRTPEELRAIKQVYEEEYGSSLEDDVVGDTSGYYQRMLVVLLQANRDPDAGIDEAQVEQDAQALFQAGELKWGTDEEKFITIFGTRSVSHLRKVFDKYMTISGFQIEETIDRETSGNLEQLLLAVVKSIRSIPAYLAETLYYAMKGAGTDDHTLIRVMVSRSEIDLFNIRKEFRKNFATSLYSMIKGDTSGDYKKALLLLCGEDDSpecifications
annexin A5 | |
Polyclonal | |
Rabbit polyclonal antibody raised against a full-length human ANXA5 protein. | |
ANXA5 | |
ANX5/ENX2/PP4 | |
Rabbit | |
Affinity Purified | |
RUO | |
308 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
Western Blot | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_001154.2 | |
ANXA5 | |
ANXA5 (NP_001145.1, 1 a.a. ∼ 320 a.a) full-length human protein. | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.