Learn More
adaptor-related protein complex 1, mu 1 subunit, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant AP1M1.
Supplier: Abnova Corporation H00008907A01
Description
The protein encoded by this gene is the medium chain of the trans-Golgi network clathrin-associated protein complex AP-1. The other components of this complex are beta-prime-adaptin, gamma-adaptin, and the small chain AP1S1. This complex is located at the Golgi vesicle and links clathrin to receptors in coated vesicles. These vesicles are involved in endocytosis and Golgi processing. Alternatively spliced transcript variants encoding distinct protein isoforms have been found for this gene. [provided by RefSeq
Sequence: MSASAVYVLDLKGKVLICRNYRGDVDMSEVEHFMPILMEKEEEGMLSPILAHGGVRFMWIKHNNLYLVATSKKNSpecifications
adaptor-related protein complex 1, mu 1 subunit | |
Polyclonal | |
Mouse polyclonal antibody raised against a partial recombinant AP1M1. | |
AP1M1 | |
AP47/CLAPM2/CLTNM/MU-1A | |
Mouse | |
50 μL | |
Yes | |
8907 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
ELISA, Western Blot | |
Unconjugated | |
50% glycerol | |
NM_032493 | |
AP1M1 | |
AP1M1 (NP_115882, 1 a.a. ∼ 74 a.a) partial recombinant protein with GST tag. | |
RUO | |
Primary | |
Human | |
Serum |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.