Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AP2 alpha Rabbit anti-Human, Mouse, Ovine, Rat, Polyclonal, Invitrogen™

Catalog No. PIPA125327
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA125327 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA125327 Supplier Thermo Scientific Supplier No. PA125327

Rabbit Polyclonal Antibody

PA1-25327 detects AP2 alpha from human samples. PA1-25327 is expected to cross react with mouse (94% conserved), rat (94% conserved), chicken (88% conserved), and ovine (94% conserved) due to sequence homology. PA1-25327 has been successfully used in Western blot and immunofluorescence procedures. The PA1-25327 immunogen is a synthetic peptide, conjugated to KLH, corresponding to residues S(420) H T D N N A K S S D K E E K H R K(437) of human AP2 alpha. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

The protein encoded by this gene is a transcription factor that binds the consensus sequence 5′-GCCNNNGGC-3′. The encoded protein functions as either a homodimer or as a heterodimer with similar family members. This protein activates the transcription of some genes while inhibiting the transcription of others. Defects in this gene are a cause of branchiooculofacial syndrome (BOFS). Three transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AP2 alpha
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Polyclonal
Concentration 0.9 mg/mL
Conjugate Unconjugated
Gene TFAP2A
Gene Accession No. P05549, P34056, Q9N0N3, P58197
Gene Alias AP-2, AP-2alpha, AP2TF, BOFS, FLJ51761, TFAP2
Gene Symbols TFAP2A
Host Species Rabbit
Immunogen Synthetic peptide, conjugated to KLH, corresponding to residues S(420) H T D N N A K S S D K E E K H R K(437) of human AP2 alpha.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21418, 306862, 443109, 7020
Target Species Human, Mouse, Ovine, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.