Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AP2B1, Mouse, Clone: 2D5, Abnova™

Catalog No. 89016061 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-016-061 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-016-061 Supplier Abnova Corporation Supplier No. H00000163M01
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant AP2B1.

The protein encoded by this gene is one of two large chain components of the assembly protein complex 2, which serves to link clathrin to receptors in coated vesicles. The encoded protein is found on the cytoplasmic face of coated vesicles in the plasma membrane. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq

Sequence: SHGIHRKHLPIHHGSTDAGDSPVGTTTATNLEQPQVIPSQGDLLGDLLNLDLGPPVNVPQVSSMQMG

Specifications

Antigen AP2B1
Applications ELISA, Immunohistochemistry (PFA fixed), Western Blot
Classification Monoclonal
Clone 2D5
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant AP2B1.
Formulation PBS with no preservative; pH 7.4
Gene AP2B1
Gene Accession No. NM_001282
Gene Alias ADTB2/AP105B/AP2-BETA/CLAPB1/DKFZp781K0743
Gene Symbols AP2B1
Host Species Mouse
Immunogen AP2B1 (NP_001273.1, 585 a.a. ∼ 651 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 163
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.