Learn More
adaptor-related protein complex 4, beta 1 subunit, Mouse, Clone: 1B2, Abnova™
Mouse monoclonal antibody raised against a partial recombinant AP4B1.
Supplier: Abnova Corporation H00010717M03
Description
The heterotetrameric adaptor protein (AP) complexes sort integral membrane proteins at various stages of the endocytic and secretory pathways. AP4 is composed of 2 large chains, beta-4 (AP4B1) and epsilon-4 (AP4E1; MIM 607244), a medium chain, mu-4 (AP4M1; MIM 602296), and a small chain, sigma-4 (AP4S1; MIM 607243).[supplied by OMIM
Sequence: VAHQQVLPWRGEFHPDTLQMALQVVNIQTIAMSRAGSRPWKAYLSAQDDTGCLFLTELLLEPGNSEMQISVKQNEARTETLNSFISVLETVIGTIEEIKSSpecifications
adaptor-related protein complex 4, beta 1 subunit | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_006594 | |
AP4B1 | |
AP4B1 (NP_006585, 640 a.a. ∼ 739 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Yes | |
10717 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG2a κ |
ELISA | |
1B2 | |
Mouse monoclonal antibody raised against a partial recombinant AP4B1. | |
AP4B1 | |
BETA-4 | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Human | |
Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.