Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AP50 Mouse, Unlabeled, Clone: 31, BD

Catalog No. BDB611351 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB611351 150 μg
BDB611350 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB611351 Supplier BD Biosciences Supplier No. 611351
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Sorting of integral membrane proteins is mediated vesicular trafficking between a variety of organelles. Two sorting signals are tyrosine-based and dileucine-based signals that interact with heterotetrameric adaptor protein complexes (AP-1, AP-2, AP-3, and AP-4), which are associated with the vesicle coats. These coatomers contain two large adaptin proteins (γ, α, δ, ε, and β1, β2, β3, β4 respectively) that are noncovalently linked to one medium chain (μ1, μ2, μ3, μ4 respectively) and one small chain ( σ1, σ2, σ3, σ4 respectively). The AP-1 and AP-3 complexes are involved in protein sorting from the TGN and endosomes, while AP-2, μ2 (AP50) interacts with integral membrane proteins via binding to tyrosine-based signals with the canonical motif YXXφ. In addition, AP50/μ2 is required for both the assembly and the proton transport activity of vacuolar (H+)-ATPases in clathrin coated vesicles. Thus, AP50/μ2 may be involved in targeting integral membrane proteins that are sorted based on tyrosine-based signals and involved in assembly of functional ion channels associated with clathrin coated vesicles.

Host Species: Mouse
Clone: 31
Isotype: IgG1
Species Reactivity: Rat
Immunogen: Mouse AP50/μ2 aa. 110-230
Formula Weight [Chemical]: 50kDa

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen AP50
Applications Western Blot
Classification Monoclonal
Clone 31
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Mouse AP50/μ2 aa. 110-230
Purification Method Affinity Purified
Quantity 150 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.