Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human APAF-1 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 8 publications
Supplier: R&D Systems MAB868
This item is not returnable.
View return policy
Description
APAF-1 Monoclonal specifically detects APAF-1 in Human samples. It is validated for Western Blot.Specifications
| APAF-1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| AAC51678 | |
| APAF1 | |
| E. coli-derived recombinant human APAF-1 Leu10-Asp214 Accession # AAC51678 | |
| 100 μg | |
| Primary | |
| Detects human APAF-1 in Western blots. | |
| Human | |
| Purified |
| Western Blot | |
| 94408 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| APAF1, APAF-1apoptotic protease activating factor 1, apoptotic peptidase activating factor, apoptotic peptidase activating factor 1, apoptotic protease activating factor, apoptotic protease-activating factor 1, CED4, DKFZp781B1145, KIAA0413 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 317 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction