Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Apaf-1 Rabbit anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma AB16503
Description
Anti-Apaf-1 Antibody, C-terminus detects level of Apaf-1 and has been published and validated for use in western blotting.
Specifications
Apaf-1 | |
Polyclonal | |
Purified IgG by immunoaffinity chromatography. Liquid in PBS containing 0.02% sodium azide. | |
Rabbit | |
100 μg | |
NM_001160.2; NM_013229.2; NM_181861.1; NM_181868.1; NM_181869.1 | |
2-8°C in undiluted aliquots for up to 12 months |
Western Blot | |
Unconjugated | |
APAF1; CED4; DKFZp781B1145; Apaf-1; KIAA0413 | |
Rabbit anti-Apaf1 (C-terminus) polyclonal antibody was raised against a peptide corresponding to amino acids 1158 to 1177 of human Apaf1 (Zou et al. 1997). The sequence of the immunogenic peptide differs from that of murine Apaf1 by one amino acid (Zou | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction