Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

APBB3, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89002824 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-002-824 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-002-824 Supplier Abnova Corporation Supplier No. H00010307B01P
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human APBB3 protein.

The protein encoded by this gene is a member of the APBB protein family. It is found in the cytoplasm and binds to the intracellular domain of the Alzheimer's disease beta-amyloid precursor protein (APP) as well as to other APP-like proteins. It is thought that the protein encoded by this gene may modulate the internalization of APP. Multiple transcript variants encoding several different isoforms have been found for this gene. [provided by RefSeq]

Sequence: MLGKDYMLAIILVNCDDDLWGDHSLEVEAGLPPGWRKIHDAAGTYYWHVPSGSTQWQRPTWELGDAEDPGTGTEGIWGLRPPKGRSFSSLESSLDRSNSLSWYGGESYIQSMEPGAKCFAVRSLGWVEVPEEDLAPGKSSIAVNNCIQQLAQTRSRSQPPDGAWGEGQNMLMILKKDAMSLVNPLDHSLIHCQPLVHIRVWGVGSSKGRDRDFAFVASDKDSCMLKCHVFRCDVPAKAIASALHGLCAQILSERVEVSGDASCCSPDPISPEDLPRQVELLDAVSQAAQKYEALYMGTLPVTKAMGMDVLNEAIGTLTARGDRNAWVPTMLSVSDSLMTAHPIQAEASTEEEPLWQCPVRLVTFIGVGRDPHTFGLIADLGRQSFQCAAFWCQPHAGGLSEAVQAACMVQYQKCLVASAARGKAWGAQARARLRLKRTSSMDSPGGPLPLPLLKGGVGGAGATPRKRGVFSLLDAFRLKPSLLHMP

Specifications

Antigen APBB3
Applications Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human APBB3 protein.
Formulation PBS with no preservative; pH 7.4
Gene APBB3
Gene Accession No. BC013158
Gene Alias FE65L2/MGC150555/MGC87674/SRA
Gene Symbols APBB3
Host Species Mouse
Immunogen APBB3 (AAH13158.1, 1 a.a. ∼ 486 a.a) full-length human protein.
Purification Method Affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10307
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.